Merck
A novel HPLC-UV/nano-TiO2-chemiluminescence system for the determination of selenocystine and selenomethionine.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-07-01)
Yingying Su, He Chen, Ying Gao, Xiaohong Li, Xiandeng Hou, Yi Lv
ABSTRAKT
Active oxygen species from the photocatalytic reaction in aqueous solution react with luminol to emit strong chemiluminescence (CL), and this can be inhibited by the UV decomposed-products of selenocystine (SeCys) or selenomethionine (SeMet). Based on this phenomenon, a novel hyphenated technique, HPLC-UV/nano-TiO(2)-CL, was established for the determination of SeCys and SeMet. The effects of pH, the UV irradiation time, the TiO(2) coated on the inner surface of the reaction tubing, and the Co(2+) catalyst concentration on the CL intensity and/or chromatographic resolution were systematically investigated. Under these optimized conditions, the inhibited CL intensity has a good linear relationship with the concentration of SeCys in the range of 0.04-10.6 microg mL(-1) or SeMet in the range of 0.05-12.4 microg mL(-1), with a limit of detection (S/N=3) of 6.4 microg L(-1) for SeCys or 12 microg L(-1) for SeMet. As an example, the method was preliminarily applied to the determination of the selenoamino acids in garlic and rabbit serum, with a recovery of 88-104%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Seleno-L-cystine, 95%
Sigma-Aldrich
Seleno-DL-cystine, powder