Merck
Conformational behavior of cinchonidine revisited: a combined theoretical and experimental study.
The journal of physical chemistry. A (2008-07-19)
Atsushi Urakawa, Daniel M Meier, Heinz Rüegger, Alfons Baiker
ABSTRAKT
Conformational space of cinchonidine has been explored by means of ab initio potential and free energy surfaces, and the temperature-induced changes of conformational populations were studied by a combined NOESY-DFT analysis. The DFT-derived potential energy surface investigation identified four new conformers. Among them, Closed(7) is substantially relevant to fully understand the conformational behavior. The energy surfaces gave access to the favored transformation pathways at different temperatures (280-320 K). They also revealed the reasons for the negligible presence of energetically stable conformers and explained the experimentally observed temperature dependence of the populations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cinchonidine, 96%