Merck
Fragrance material review on sclareol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
S P Bhatia, D McGinty, C S Letizia, A M Api
ABSTRAKT
A toxicologic and dermatologic review of sclareol when used as a fragrance ingredient is presented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sclareol, 98%