Merck
Fragrance material review on cedrol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
S P Bhatia, D McGinty, C S Letizia, A M Api
ABSTRAKT
A toxicologic and dermatologic review of cedrol when used as a fragrance ingredient is presented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(+)-Cedrol, ≥99.0% (sum of enantiomers, GC)