Merck
[Synthesis of diethyl N-[p-(methylamino)benzoyl]-L-glutamate].
Beijing da xue xue bao. Yi xue ban = Journal of Peking University. Health sciences (2008-08-05)
Chao Tian, Shou-xin Zhou, Biao Wang, Xi-ling Deng, Ying Guo, Zhi-li Zhang, Xiao-wei Wang, Jun-yi Liu
ABSTRAKT
To find a best synthesis method of diethyl N-[p-(methylamino)benzoyl]-L-glutamate(1). Using diethyl N-(p- aminobenzoyl)-L-glutamate (2) as a starting material via a one-pot process of benzylation and methylation, the diethyl N-[p-(methylamino)benzoyl ]-L-glutamate (3) is prepared in high yield. Then compound 1 was conveniently obtained from debenzylation of compound 3 by Pd/C. The target compound was obtained through 2-step reaction in 88% overall yield in only 6 h, and its structure was identified by 1HNMR and MS. It is a convenient and efficient method of preparation of diethyl N-[p-(methylamino)benzoyl]-L-glutamate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Glutamic acid diethyl ester hydrochloride, 97%