- Identification of 14-3-3zeta by chemical affinity with salicylanilide inhibitors of interleukin-12p40 production.
Identification of 14-3-3zeta by chemical affinity with salicylanilide inhibitors of interleukin-12p40 production.
Bioconjugate chemistry (2008-09-04)
Jim P Boyce, Michael E Brown, Wilson Chin, Jeffrey N Fitzner, Raymond J Paxton, Min Shen, Tracey Stevens, Martin F Wolfson, Clifford D Wright
PMID18763820
ABSTRAKT
Salicylanilides were found as selective inhibitors of interleukin-12p40 production in stimulated dendritic cells. The conversion of one of these bioactive salicylanilides into a comparably bioactive, chemically labeled derivative was achieved using a facile and systematic functional group derivatization strategy. This resulted in a tool reagent that was then employed in an affinity chromatography approach that resulted in the identification of the protein 14-3-3zeta as having selective affinity for the chromatography matrix that was derivatized with a salicylanilide that inhibited IL-12p40 production.