Merck
Antioxidant activity of a combinatorial library of emulsifier-antioxidant bioconjugates.
Journal of agricultural and food chemistry (2008-09-12)
Charlotte S Hunneche, Marianne N Lund, Leif H Skibsted, John Nielsen
ABSTRAKT
A combinatorial chemistry approach was employed for the design and systematic synthesis of antioxidant-emulsifier bioconjugates to improve antioxidant activity in the interface between oil and water. A combinatorial library of 12 bioconjugates was synthesized from the phenolic antioxidants Trolox (a water-soluble alpha-tocopherol analogue), dihydroferulic acid, dihydrocaffeic acid, and gallic acid in combination with serine ethyl ester, serine lauryl ester, and lauroyl serine by esterification of the serine side chain or amidation, respectively. The bioconjugates were characterized by NMR and mass spectrometry, and each was tested for antioxidant activity by measuring the radical scavenging rate of 2,2-diphenyl-1-picrylhydrazyl (DPPH (*)) in methanol, the radical scavenging rate of DPPH (*) in a heterogeneous solvent system, the rate of oxygen consumption in an oil-in-water emulsion with metmyoglobin initiated oxidation, and the lag phase for diene formation in unilamellar liposomes with free radical initiation in the aqueous phase; each case was compared to the antioxidant activity of the parent antioxidant. In general, the conjugates with longer chain derivatives exhibited improved antioxidative activity in heterogeneous systems, whereas no improvement was observed in homogeneous solution. The rate of oxygen consumption in oil-in-water emulsion was found to decrease with increasing octanol/water partition coefficient, which is suggested to correspond to a saturation of the water/oil interface with antioxidant bioconjugate to approach maximal protection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%