Merck
Determination of trihalomethanes in waters by ionic liquid-based single drop microextraction/gas chromatographic/mass spectrometry.
Journal of chromatography. A (2008-09-27)
Eva Aguilera-Herrador, Rafael Lucena, Soledad Cárdenas, Miguel Valcárcel
ABSTRAKT
A simple, rapid, solventless method for the determination of trihalomethanes (THMs) (chloroform, bromodichloromethane, dibromochloromethane and bromoform) in water samples is presented. The analytes are extracted from the headspace of the aqueous matrix into a 2 microL drop of the ionic liquid 1-octyl-3-methyl-imidazolium hexafluorophosphate working at 30 degrees C for 30 min. The separation and detection of the target compounds is accomplished by gas chromatography/mass spectrometry owing to the use of an interface that efficiently transfers the analytes extracted in the ionic liquid drop to the gas chromatograph while preventing the ionic liquid from entering the column. The detection limits obtained are below the values compelled by the legislation, ranging from 0.5 microg L(-1) for chloroform and bromodichloromethane to 0.9 microg L(-1) for dibromochloromethane. The use of ionic liquid in the extraction procedure avoids the use of organic solvents and leads to relative standard deviations that range from 3.1% to 4.8%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Bromoform solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Bromodichloromethane, ≥97%
Sigma-Aldrich
Dibromochloromethane, 97%
Sigma-Aldrich
Bromoform, contains 60-120 ppm 2-methyl-2-butene as stabilizer, 99%
Sigma-Aldrich
Bromoform, contains 1-3% ethanol as stabilizer, 96%
Supelco
Bromoform, amylene stabilized, analytical standard