Merck
Chiral squaramide derivatives are excellent hydrogen bond donor catalysts.
Journal of the American Chemical Society (2008-10-14)
Jeremiah P Malerich, Koji Hagihara, Viresh H Rawal
ABSTRAKT
Thioureas represent the dominant platform for hydrogen bond promoted asymmetric catalysts. A large number of reactions, reported in scores of publications, have been successfully promoted by chiral thioureas. The present paper reports the use of squaramides as a highly effective new scaffold for the development of chiral hydrogen bond donor catalysts. Squaramide catalysts are very simple to prepare. The (-)-cinchonine modified squaramide (5), easily prepared through a two-step process from methyl squarate, was shown to be an effective catalyst, even at catalyst loadings as low as 0.1 mol%, for the conjugate addition reactions of 1,3-dicarbonyl compounds to beta-nitrostyrenes. The addition products were obtained in high yields and excellent enantioselectivities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cinchonine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
3,4-Dihydroxy-3-cyclobutene-1,2-dione, ≥99.0% (HPLC)
Sigma-Aldrich
(+)-Cinchonine, 85%
Sigma-Aldrich
trans-β-Nitrostyrene, 99%