Merck
Fluorometric determination of carbohydrate with 2-aminothiophenol.
Analytical biochemistry (1991-05-15)
J K Zhu, E A Nothnagel
ABSTRAKT
The 2-aminothiophenol-based fluorometric assay of Nakano et al. (1973, J. Pharm. Soc. Jpn. 93, 350-353) for monosaccharides has been modified to improve the speed, applicability, and sensitivity of the method. The improved assay is applicable to complex carbohydrates as well as to monosaccharides. Less than 50 ng of carbohydrate in a final volume of 2 ml can be quantitatively measured within 30 min. The assay is reasonably compatible with the presence of a variety of reagents commonly used in aqueous buffer solutions. The assay is especially useful for monitoring column eluents during the purification of small quantities of carbohydrates or their conjugates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Aminothiophenol, technical grade, 90%
Sigma-Aldrich
2-Aminothiophenol, 99%