Merck
Effects of preparation and experimental conditions on removal of phenol by surfactant-modified zeolites.
Environmental technology (2008-11-04)
C Díaz-Nava, M T Olguín, M Solache-Ríos, M T Alarcón-Herrera, A Aguilar-Elguezabal
ABSTRAKT
The removal of phenol from aqueous solutions using surfactant-modified clinoptilolite-rich tuffs (SMZ) prepared from two Mexican zeolitic tuffs (Chihuahua and Oaxaca) was investigated. Sodium homoionization of the zeolitic rocks was performed before they were modified with the surfactants: hexadecyltrimethylammonium chloride or bromide and bencylcetildimethylammonium chloride. The surfactants in the modified zeolitic materials were determined considering the total carbon in the samples. The phenol removal was determined in a batch system taking into consideration the different quantities of surfactants in the zeolitic tuffs, contact time, pH and initial phenol concentration. The phenol was determined in the aqueous solutions by UV/Vis spectroscopy. Results showed that the formation of a hemimicelle or admicelle on the zeolites depended on the initial surfactant concentration and they were responsible for the type of interactions between the phenol and the surfactant-modified zeolites. Phenol adsorption by the surfactant-modified zeolites was carried out in approximately three hours. Phenol adsorption data was best adjusted to the pseudo-second order kinetic model. Both, surface properties of the surfactant-modified zeolites and pH of solution play an important role in the removal of this pollutant from aqueous solutions and they are responsible for the type of mechanism involved.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Benzyldimethylhexadecylammonium chloride, cationic detergent