Merck
Nickel-catalyzed decarbonylative addition of anhydrides to alkynes.
Journal of the American Chemical Society (2008-12-04)
Yuichi Kajita, Takuya Kurahashi, Seijiro Matsubara
ABSTRAKT
An intermolecular nickel-catalyzed addition reaction has been developed where phthalic anhydrides react with alkynes to afford substituted isocoumarins. A mechanistic rationale is proposed, implying reductive elimination of Ni(0) promoted by ZnCl(2) cocatalyst as the key step of the catalytic cycle.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phthalic anhydride, ACS reagent, ≥99%
Sigma-Aldrich
Phthalic anhydride, ReagentPlus®, 99%