Merck
  • Validated HPLC method for the standardization of Phyllanthus niruri (herb and commercial extracts) using corilagin as a phytochemical marker.
Validated HPLC method for the standardization of Phyllanthus niruri (herb and commercial extracts) using corilagin as a phytochemical marker.
Biomedical chromatography : BMC (2009-03-12)
Renata Colombo, Andrea N de L Batista, Helder L Teles, Geraldo H Silva, Giovani C C Bomfim, Rosilene C R Burgos, Alberto J Cavalheiro, Vanderlan da Silva Bolzani, Dulce Helena S Silva, Clícia R Pelícia, Fabiana M Guimarães, Maria Célia H Heimberg
ABSTRAKT
Phyllanthus niruri L., commonly known in Brazil as 'quebra-pedra', has long been used in the treatment of diverse diseases and especially urolithiasis. The therapeutic effects of P. niruri are attributed to various compounds present in the plant, including the hydrolysable tannin corilagin. In the present study, high-performance liquid chromatography (HPLC-/PAD) profiles of leaves and commercial extracts of P. niruri were examined and three compounds, found to be present in all of the samples studied, were isolated by open column chromatography over C18)silica gel followed by preparative HPLC. These compounds were identified by nuclear magnetic resonance as corilagin, rutin and ethyl 3,4,5-trihydroxybenzoate. Corilagin, which has been proposed as a phytochemical marker for P. niruri, was employed as an external standard in the development and validation of a rapid and efficient qualitative and quantitative HPLC assay for the analyte. The method may be applied in the standardization of herbs and phytomedicines commercialized in Brazil as quebra-pedra.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Corilagin
Supelco
Corilagin, analytical standard
Sigma-Aldrich
Ethyl gallate, antioxidant, ≥96.0% (HPLC)