Merck
Occurrence of dissolved and particle-bound taste and odor compounds in Swiss lake waters.
Water research (2009-03-24)
Andreas Peter, Oliver Köster, Andrea Schildknecht, Urs von Gunten
ABSTRAKT
The occurrence of algal taste and odor (T&O) compounds was investigated in three Swiss lakes which exhibit different nutrient levels from eutrophic to oligotrophic (Lake Greifensee, Lake Zurich and Lake Lucerne). Apart from dissolved T&O compounds, the study also encompassed particle-bound compounds, i.e., compounds that can be released from damaged algal cells during drinking water treatment. A combined instrumental (SPME-GC-MS) and sensory method was applied that allowed to detect and quantify T&O compounds in natural waters in the sub ppt to low ppt-range. In addition to the prominent T&O compounds geosmin and 2-methyl-isoborneol (MIB), four other T&O compounds could be detected in the lake waters, though all at relatively low concentrations (maximum concentrations of geosmin 19 ng L(-1), MIB 3 ng L(-1), beta-ionone 27 ng L(-1), beta-cyclocitral 7 ng L(-1), 2-isobutyl-3-methoxypyrazine 2 ng L(-1), 2-isopropyl-3-methoxypyrazine 16 ng L(-1)). The concentration peaks typically occurred in the epilimnion during summer concurrent with a high phytoplankton biomass. Consistently, the concentration levels for most of the compounds varied substantially between the three lakes and generally decreased in the order eutrophic Lake Greifensee>mesotrophic Lake Zurich>oligotrophic Lake Lucerne. Furthermore, our data revealed that the occurrence of beta-ionone was largely influenced by Planktothrix rubescens. This is the first time that a correlation between beta-ionone and this cyanobacterium has been reported for natural waters.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Isobutyl-3-methoxypyrazine, 99%
Supelco
2-Isobutyl-3-methoxypyrazine solution, certified reference material
Sigma-Aldrich
2-Isobutyl-3-methoxypyrazine, ≥99%, FG
Supelco
2-Isopropyl-3-methoxypyrazine solution, certified reference material