Merck
Enzyme-enhanced fluorescence detection of DNA on etched optical fibers.
Biosensors & bioelectronics (2009-03-25)
Shu-yan Niu, Quan-yi Li, Rui Ren, Shu-sheng Zhang
ABSTRAKT
A novel DNA biosensor based on enzyme-enhanced fluorescence detection on etched optical fibers was developed. The hybridization complex of DNA probe and biotinylated target was formed on the etched optical fiber, and was then bound with streptavidin labeled horseradish peroxidase (streptavidin-HRP). The target DNA was quantified through the fluorescent detection of bi-p,p'-4-hydroxyphenylacetic acid (DBDA) generated from the substrate 4-hydroxyphenylacetic acid (p-HPA) under the catalysis of HRP, with a detection limit of 1 pM and a linear range from 1.69 pM to 169 pM. It is facile to regenerate this sensor through surface treatment with concentrated urea solution. It was discovered that the sensor can retain 70% of its original activity after three detection-regeneration cycles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%