Merck
Microcystins -LA, -YR, and -LR action on neutrophil migration.
Biochemical and biophysical research communications (2009-04-11)
Paula Kujbida, Elaine Hatanaka, Marco Aurélio Ramirez Vinolo, Kaline Waismam, Danielle Maia de Holanda Cavalcanti, Rui Curi, Sandra Helena Poliselli Farsky, Ernani Pinto
ABSTRAKT
Microcystins (MCs) produced by some freshwater cyanobacterial species possess potent liver toxicity as evidenced by acute neutrophil infiltration. Here, we investigate the ability of three structurally distinct toxins (MC-LA, MC-LR, and MC-YR) to evoke neutrophil recruitment per se and their effects on migration pathways. Intravital microscopic studies showed that topical application of only MC-LR enhanced the numbers of rolling and adhered leukocytes in the endothelium of postcapillary mesenteric venules. The latter effects may be dependent upon induction of the synthesis and expression of L-selectin and beta2-integrin in neutrophils, as assessed by flow cytometry and RT-PCR, respectively. Conversely, the three toxins promoted direct locomotion of neutrophils and enhanced their migration in response to fMLP, as measured by Boyden chamber assays, and increased intracellular calcium, a messenger in the chemotaxic process. In conclusion, our results show that MCs act on specific pathways of neutrophil recruitment, indicating their potential effect on neutrophils activation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Microcystin-YR solution, 10 μg/mL in methanol, analytical standard