Merck
CuBr-catalyzed oxidative difluoromethylation of tertiary amines with difluoroenol silyl ethers.
Organic letters (2009-04-22)
Lingling Chu, Xingang Zhang, Feng-Ling Qing
ABSTRAKT
CuBr-catalyzed oxidative difluoromethylation of readily available tertiary amines with difluoroenol silyl ethers was performed under mild conditions to afford beta-amine-alpha,alpha-difluoro ketones.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Copper(II) bromide, 99%
Sigma-Aldrich
Copper(II) bromide, 99.999% trace metals basis