Merck
Deprotection of N(in)-formyl group on tryptophan by application of new reagent, 1,2-dimethylethylendiamine in aqueous solution.
Advances in experimental medicine and biology (2009-04-30)
Takenao Odagami, Yuji Kogami, Hiroyuki Kouji, Yuko Tsuda, Yoshio Okada
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 85%
Sigma-Aldrich
N,N′-Dimethylethylenediamine, 98%