Merck
Influence of structural variations on drug release from lipid/polyethylene glycol matrices.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2009-05-02)
Maike Windbergs, Clare Joanna Strachan, Peter Kleinebudde
ABSTRAKT
Different combinations of monoacid triglycerides and polyethylene glycol powders of different molecular weights were successfully extruded below their melting temperatures as a basis for oral dosage forms. The incorporated polyethylene glycols exhibiting different mean molecular weights were compared with respect to their effect on the dissolution rate for a model drug, and a mean molecular weight of 10,000 resulted in the most advantageous characteristics (fastest dissolution). Triglycerides with different fatty acid chain lengths provided additional options for the extrusion temperature which is beneficial for temperature sensitive active pharmaceutical ingredients. To obtain stable extrudates with desirable and reproducible dissolution characteristics a good understanding of the underlying processes during processing and storage is mandatory.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glyceryl tridodecanoate, ≥99%
Sigma-Aldrich
Glyceryl tristearate, ≥99%
Sigma-Aldrich
Glyceryl tristearate, technical
Sigma-Aldrich
Glyceryl tripalmitate, ≥85%
Sigma-Aldrich
Glyceryl tripalmitate, ≥99%