Merck
Nutmeg (Myristica fragrans Houtt.).
Disease-a-month : DM (2009-05-19)
Donald G Barceloux
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Myristicin from parsley leaf oil, ≥85% (HPLC), oil
Supelco
Myristicin, analytical standard