Merck
Lactosylceramide promotes cell migration and proliferation through activation of ERK1/2 in human aortic smooth muscle cells.
American journal of physiology. Heart and circulatory physiology (2009-05-26)
Hong Mu, Xinwen Wang, Hao Wang, Peter Lin, Qizhi Yao, Changyi Chen
ABSTRAKT
Increased plasma levels of lactosylceramide (LacCer) have been associated with cardiovascular disease. However, it is largely unknown whether LacCer directly contributes to dysfunction of smooth muscle cells (SMCs), a key event in vascular lesion formation. In the present study, we determined the effects and potential mechanisms of LacCer on cell migration and proliferation in human aortic SMCs (AoSMCs). Cell migration and proliferation were determined by a modified Boyden chamber assay and nonradioactive colorimetric (MTS) assay, respectively. We found that LacCer significantly induced AoSMC migration and proliferation in a concentration- and time-dependent manner. In addition, LacCer significantly upregulated the expression of PDGFR-B, integrins (alpha(v) and beta(3)), and matrix metalloproteinases (matrix metalloproteinase-1 and -2) at both mRNA and protein levels, as determined by real-time PCR and Western blot analyses, respectively. Furthermore, LacCer increased superoxide anion production and the transient phosphorylation of ERK1/2 in AoSMCs, as determined by dihydroethidium staining and immunoassay, respectively. Accordingly, LacCer-induced cell migration and proliferation were effectively blocked by antioxidants (seleno-l-methionine and Mn tetrakis porphyrin) and by a specific ERK1/2 inhibitor. Thus, LacCer promotes cell migration and proliferation through oxidative stress and activation of ERK1/2 in AoSMCs. These findings demonstrate the functional role of LacCer in the vascular disease pathogenesis.