Merck
  • Persistent organic pollutants in fish oil supplements on the canadian market: polychlorinated dibenzo-p-dioxins, dibenzofurans, and polybrominated diphenyl ethers.
Persistent organic pollutants in fish oil supplements on the canadian market: polychlorinated dibenzo-p-dioxins, dibenzofurans, and polybrominated diphenyl ethers.
Journal of food science (2009-06-06)
D F K Rawn, K Breakell, V Verigin, H Nicolidakis, D Sit, M Feeley, J J Ryan
ABSTRAKT
Canadians are interested in improving their diet through the consumption of fish oil food supplements, which are marketed to be rich in omega-3 fatty acids. Convenience samples of omega-3 enriched dietary supplements (n = 30) were collected in Vancouver, Canada, between 2005 and 2007. All of the omega-3 supplements were analyzed for polychlorinated dibenzo-p-dioxins/furans (PCDD/Fs) and polybrominated diphenyl ethers (PBDEs) and, although every sample was found to contain detectable residues of PBDEs, only 24 samples were found to have PCDD/F concentrations above the level of detection. PCDD/F concentrations ranged from 0.05 pg TEQ/g lipid to 45.7 pg TEQ/g lipid in salmon and shark oils, respectively. Maximum PBDE concentrations similarly were observed in shark oil (113 microg/kg lipid), however, most supplements had concentrations below 5 microg/kg lipid. Average PCDD/F and PBDE intake estimates, based on consumption of maximum supplement dose following product label recommendations, were 4.32 pg TEQ/d and 25.1 ng/d lipid, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fish oil from menhaden, crude source of omega-3 fatty acids