Merck
Molecularly imprinted tunable binding sites based on conjugated prosthetic groups and ion-paired cofactors.
Journal of the American Chemical Society (2009-06-06)
Kohei Takeda, Atsushi Kuwahara, Kohei Ohmori, Toshifumi Takeuchi
ABSTRAKT
A molecular imprinting technique was applied to the construction of prosthetic group-coupled tunable binding cavities for bisphenol A (BPA). A novel template molecule, with a structure consisting of BPA covalently conjugated with two allyl(4-carboxyphenyl)disulfides through ester bonds (BPA-D), was designed. After copolymerization of BPA-D with styrene and divinylbenzene, the BPA di(4-mercaptobenzoate) moieties were removed by reductive cleavage of the disulfide bonds, resulting in apo-type molecularly imprinted cavities bearing two thiol residues. 4-Mercaptobenzoic acid was introduced into the apo-type cavities as a prosthetic group through a disulfide bond by addition of 4,4'-dithiodibenzoic acid, which transformed the apo-type cavities into holo-type cavities with two carboxylic acid residues for binding BPA. When pyridyl prosthetic groups were introduced instead of 4-mercaptobenzoic acid by using 4,4'-dithiodipyridine, BPA recognition ability was maintained but with improved selectivity. The binding affinity was successfully altered several times by attaching and detaching these prosthetic groups, which showed that the apo-type scaffold could be reused. Furthermore, noncovalent-type ion-paired cofactors could be introduced, when the two thiol groups in the apo-type cavities were oxidized to sulfonic acid groups. When 1,2-diaminoethane (DAE) was added to the oxidized apo-type scaffold as a noncovalent-type cofactor, the binding activity was regulated successively, depending upon the concentrations of DAE added. By using various prosthetic groups and cofactors, the binding properties of the holo-type cavities could be tuned in a similar way to those found in biological systems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Mercaptobenzoic acid, 99%
Sigma-Aldrich
4-Mercaptobenzoic acid, technical grade, 90%
Sigma-Aldrich
Aldrithiol-4, 98%