Merck
  • Temperature-induced inversion of elution order in the chromatographic enantioseparation of 1,1'-bi-2-naphthol on an immobilized polysaccharide-based chiral stationary phase.
Temperature-induced inversion of elution order in the chromatographic enantioseparation of 1,1'-bi-2-naphthol on an immobilized polysaccharide-based chiral stationary phase.
Journal of chromatography. A (2009-06-09)
Bixia Yao, Fengping Zhan, Guangyan Yu, Zhifen Chen, Wenjing Fan, Xiongping Zeng, Qingle Zeng, Wen Weng
ABSTRAKT
In this work, the enantioseparations of 1,1'-bi-2-naphthol (BINOL) and its three derivatives were performed on an immobilized polysaccharide-based chiral stationary phase, Chiralpak IA, under normal-phase mode. The effects of the content of polar modifier in the mobile phase and the column temperature on the retention and enantioseparation were investigated in detail. Temperature-induced inversion of elution order for BINOL was observed directly when n-hexane/2-propanol (92/8, v/v) was used as mobile phase. The isoenantioselective temperature (T(iso)) was calculated to be 31.4 degrees C. When n-hexane/2-propanol/THF (93/2/5, v/v/v) was used as mobile phase, the T(iso) value decreased to -8.2 degrees C. Entropically driven enantioseparation which had practical application was obtained successfully (separation factor being 1.189 and 1.332 at 25 degrees C and 50 degrees C, respectively). The corresponding thermodynamic parameters for other three binaphthyl compounds were compared with that for BINOL. Some inferences about chiral recognition mechanism were stressed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,1′-Bi-2-naphthol, 99%