Merck
  • Asymmetric synthesis of 4H-1,3-oxazines: enantioselective reductive cyclization of N-acylated beta-amino enones with trichlorosilane catalyzed by chiral Lewis bases.
Asymmetric synthesis of 4H-1,3-oxazines: enantioselective reductive cyclization of N-acylated beta-amino enones with trichlorosilane catalyzed by chiral Lewis bases.
Chemical communications (Cambridge, England) (2009-06-13)
Masaharu Sugiura, Mako Kumahara, Makoto Nakajima
ABSTRAKT
N-Acylated beta-amino enones reductively cyclize by treatment with trichlorosilane and a chiral Lewis base catalyst to afford optically active 4H-1,3-oxazines, which can be transformed to other chiral compounds without racemization.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trichlorosilane, 99%