Merck
Dequalinium, a new inhibitor of Mycobacterium tuberculosis mycothiol ligase identified by high-throughput screening.
Journal of biomolecular screening (2009-06-16)
Maria-Teresa Gutierrez-Lugo, Heather Baker, Joseph Shiloach, Helena Boshoff, Carole A Bewley
ABSTRAKT
Mycothiol ligase (MshC) is a key enzyme in the biosynthesis of mycothiol, a small molecular weight thiol that is unique to actinomycetes and whose primary role is to maintain intracellular redox balance and remove toxins. MshC catalyzes the adenosine triphosphate (ATP)-dependent condensation of cysteine and glucosamine-inositol (GI) to produce cysteine-glucosamine-inositol (CGI). MshC is essential to Mycobacterium tuberculosis and therefore represents an attractive target for chemotherapeutic intervention. A screening protocol was developed to identify MshC inhibitors based on quantification of residual ATP using a coupled luminescent assay. The protocol was used to screen a library of 3100 compounds in a 384-well plate format (Z'>or=0.65). Fifteen hits (0.48%) were identified from the screen, and 2 hits were confirmed in a secondary assay that measures production of CGI. The structures of both hits contain N-substituted quinolinium moieties, and the more potent of the 2-namely, dequalinium chloride-inhibits MshC with an IC50 value of 24+/-1 microM. Further studies showed dequalinium to be an ATP-competitive inhibitor of MshC, to bind MshC with a KD of 0.22 microM, and to inhibit the growth of M. tuberculosis under aerobic and anaerobic conditions with minimum inhibitory and anaerobic bactericidal concentrations of 1.2 and 0.3 microg/mL, respectively. The screening protocol described is robust and has enabled the identification of new MshC inhibitors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Dequalinium chloride, Pharmaceutical Secondary Standard; Certified Reference Material