Merck
Synthesis and self-assembly of amphiphilic hyperbranched polyglycerols modified with palmitoyl chloride.
Journal of colloid and interface science (2009-06-23)
Haixing Cheng, Siguang Wang, Jintian Yang, Yongfeng Zhou, Deyue Yan
ABSTRAKT
A series of palmitoyl chloride-grafted hyperbranched polyglycerols were synthesized through cationic ring-opening polymerization and end-group modification. The obtained grafted copolymer of HPG-C16 possesses a hydrophilic hyperbranched polyether core and many hydrophobic alkyl arms, which shows interesting self-assembly behavior in THF and water. The results indicate that the copolymers with a relatively high alkyl grafting ratio can form unimolecular micelles in THF and will further assemble into giant vesicles around 1-10 microm in THF/water mixed solvents. However, the HPG-C16 with a low alkyl grafting ratio of 15.6% can directly assemble into vesicles in THF and form micelles in water.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Palmitoyl chloride, 98%