Merck
Determination of Prometryne in water and soil by HPLC-UV using cloud-point extraction.
Talanta (2009-06-30)
Jihai Zhou, Jiandong Chen, Yanhong Cheng, Daming Li, Feng Hu, Huixin Li
ABSTRAKT
A CPE-HPLC (UV) method has been developed for the determination of Prometryne. In this method, non-ionic surfactant Triton X-114 was first used to extract and pre-concentrate Prometryne from water and soil samples. The separation and determination of Prometryne were then carried out in an HPLC-UV system with isocratic elution using a detector set at 254 nm wavelength. The parameters and variables that affected the extraction were also investigated and the optimal conditions were found to be 0.5% of Triton X-114 (w/v), 3% of NaCl (w/v) and heat-assisted at 50 degrees C for 30 min. Using these conditions, the recovery rates of Prometryne ranged from 92.84% to 99.23% in water and 85.48% to 93.67% in soil, respectively, with all the relative standard deviations less than 3.05%. Limit of detection (LOD) and limit of quantification (LOQ) were 3.5 microg L(-1) and 11.0 microg L(-1) in water and 4.0 microg kg(-1) and 13.0 microg kg(-1) in soil, respectively. Thus, we developed a method that has proven to be an efficient, green, rapid and inexpensive approach for extraction and determination of Prometryne from soil samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Prometryn, PESTANAL®, analytical standard