Merck
Zeolite-catalyzed isomerization of triose sugars.
ChemSusChem (2009-06-30)
Esben Taarning, Shunmugavel Saravanamurugan, Martin Spangsberg Holm, Jianmin Xiong, Ryan M West, Claus Hviid Christensen
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyl DL-lactate, ≥97.0% (GC)
Sigma-Aldrich
(+)-Methyl D-lactate, 98%, optical purity ee: 96% (GLC)
Sigma-Aldrich
(−)-Methyl L-lactate, 98%, optical purity ee: 97% (GLC)