Merck
Unexpected fluorescent behavior of a 4-amino-1,8-naphthalimide derived beta-cyclodextrin: conformation analysis and sensing properties.
Chemical communications (Cambridge, England) (2009-07-02)
Cheng Zhong, Tong Mu, Lei Wang, Enqin Fu, Jingui Qin
ABSTRAKT
A new fluorescent cyclodextrin shows unexpected and strong fluorescence enhancement upon binding of organic molecules, and the enhancing mechanism is found to be different from those reported in the literature.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Amino-1,8-naphthalimide
Sigma-Aldrich
1-Adamantanecarboxylic acid, 99%