Merck
Aldehyde selective Wacker oxidations of phthalimide protected allylic amines: a new catalytic route to beta3-amino acids.
Journal of the American Chemical Society (2009-07-09)
Barbara Weiner, Alejandro Baeza, Thomas Jerphagnon, Ben L Feringa
ABSTRAKT
A new method for the synthesis of beta(3)-amino acids is presented. Phthalimide protected allylic amines are oxidized under Wacker conditions selectively to aldehydes using PdCl(2) and CuCl or Pd(MeCN)(2)Cl(NO(2)) and CuCl(2) as complementary catalyst systems. The aldehydes are produced in excellent yields and exhibit a large substrate scope. Beta-amino acids and alcohols are synthesized by oxidation or reduction and subsequent deprotection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phthalimide, ≥99%
Sigma-Aldrich
Phthalimide potassium salt, 98%
Sigma-Aldrich
Phthalimide potassium salt, purum, ≥99.0% (NT)