Merck
Acidic-store depletion is required for human platelet aggregation.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis (2009-07-10)
Nidhal Ben Amor, Hanene Zbidi, Aicha Bouaziz, Isaac Jardin, Jardin Isaac, Juan M Hernández-Cruz, Ginés M Salido, Juan A Rosado, Aghleb Bartegi
ABSTRAKT
Platelet stimulation with thrombin induces an elevation in cytoplasmic free Ca(2+) concentration ([Ca(2+)]c) due to Ca(2+) release from intracellular stores and entry from the extracellular medium. Two different intracellular Ca(2+) stores have been described in human platelets: the dense tubular system and the lysosomal-like acidic stores. In the present study, we investigated the contribution of the acidic stores in thrombin-induced platelet aggregation. We have found that platelet aggregation induced by thrombin is reduced in a Ca(2+)-free medium. Discharge of the acidic Ca(2+) stores by treatment with the sarcoendoplasmic Ca(2+)-ATPase (SERCA)3 selective inhibitor 2,5-di-(tert-butyl)-1,4-hydroquinone reduced thrombin-evoked platelet aggregation. In the presence of 2,5-di-(tert-butyl)-1,4-hydroquinone, platelet aggregation induced by the protease-activated receptor (PAR)-1 and PAR-4 agonist peptides, SFLLRN and AYPGKF, respectively, was significantly reduced. In cells with depleted acidic stores, activation of GPIb-IX-V by thrombin resulted in reduced or no platelet aggregation in a medium containing 1 mmol/l Caor in a Ca(2+)-free medium, respectively. This finding suggests that Ca(2+) accumulation in the acidic Ca(2+) compartments is required for platelet aggregation induced by activation of the G-coupled PAR-1 and PAR-4 thrombin receptors and, by the occupation of the leucine-rich glycoprotein GPIb-IX-V and provide evidence supporting a functional role of the lysosomal-like acidic Ca(2+) stores in human platelets.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,5-Di-tert-butylhydroquinone, 99%