- Dihydropyridine C-glycoconjugates by organocatalytic Hantzsch cyclocondensation. Stereoselective synthesis of alpha-threofuranose C-nucleoside enantiomers.
Dihydropyridine C-glycoconjugates by organocatalytic Hantzsch cyclocondensation. Stereoselective synthesis of alpha-threofuranose C-nucleoside enantiomers.
Organic & biomolecular chemistry (2009-07-11)
Diogo R B Ducatti, Alessandro Massi, Miguel D Noseda, Maria Eugênia R Duarte, Alessandro Dondoni
PMID19590795
ABSTRAKT
The Hantzsch reaction of C-glycosyl aldehyde/enamino ester/beta-ketoester systems under l-proline catalysis to give dihydropyridine C-glycoconjugates is reported. Asymmetric cyclocondensations of differentially substituted enamine and beta-dicarbonyl components with formyl alpha-L-C-threofuranoside and with the alpha-D-isomer were also carried out. Each reaction occurred with high yet opposite stereoselectivity (de >95%) so that the pair of alpha-threofuranose C-nucleoside enantiomers was prepared.