Merck
[Studies on chemical constituents from roots of Peucedanum praeruptorum III].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2009-07-31)
Cun Zhang, Yongqing Xiao, Massahiko Taniguchi, Kimiye Baba
ABSTRAKT
To study the chemical constituents in roots of Peucedanum praeruptorum. The constituents were isolated by column chromatography on silica gel and ODS, and identified by spectroscopic methods. Seven compounds, alpha-D-glucopyranose-1-hexadecanoate (1), D-mannitol monohexadecanoate (2), adenosine (3), butyric acid (4), eleutheroside B, (5), apiosylskimmin (6), and mannitol (7) were isolated and identified. Compounds 1-5 were isolated from this plant for the first time.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Eleutheroside B, analytical standard