Merck
Mass attenuation coefficient of chromium and manganese compounds around absorption edge.
Journal of X-ray science and technology (2009-08-01)
Sharanabasappa, S B Kaginelli, B R Kerur, S Anilkumar, B Hanumaiah
ABSTRAKT
The total mass attenuation coefficient for Potassium dichromate, Potassium chromate and Manganese acetate compounds are measured at different photon energies 5.895, 6.404, 6.490, 7.058, 8.041 and 14.390 keV using Fe-55, Co-57 and 241Am source with Copper target, radioactive sources. The photon intensity is analyzed using a high resolution HPGe detector system coupled to MCA under good geometrical arrangement. The obtained values of mass attenuation coefficient values are compared with theoretical values. This study suggests that measured mass attenuation coefficient values at and near absorption edges differ from the theoretical value by about 5-28%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium chromate, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium chromate, ACS reagent, ≥99.0%