- Rh-catalyzed reductive coupling reaction of aldehydes with conjugated dienes promoted by triethylborane.
Rh-catalyzed reductive coupling reaction of aldehydes with conjugated dienes promoted by triethylborane.
Organic letters (2009-08-12)
Masanari Kimura, Daisuke Nojiri, Masahiro Fukushima, Shuichi Oi, Yusuke Sonoda, Yoshio Inoue
PMID19663391
ABSTRAKT
Rh(I) catalyzes the reductive coupling reaction of a wide variety of aldehydes with conjugated dienes in the presence of a stoichiometric amount of triethylborane to provide homoallyl alcohols in a single operation.