Merck
Inhibition of IgE-induced mast cell activation by ethyl tertiary-butyl ether, a bioethanol-derived fuel oxygenate.
The Journal of pharmacy and pharmacology (2009-08-26)
Kouya Yamaki, Shin Yoshino
ABSTRAKT
The effect of ethyl tertiary-butyl ether (ETBE), which is widely used as a fuel oxygenate commonly produced from bioethanol, on immunoglobulin (Ig)E-dependent mast cell activation was investigated. The rat mast cell line RBL2H3 sensitised with monoclonal anti-ovalbumin IgE was challenged with ovalbumin in the presence or absence of ETBE, tert-butanol (TBA), which is the main metabolite of ETBE in humans, and ethanol. Degranulation of RBL2H3 was examined by the release of beta-hexosaminidase. To understand the mechanisms responsible for regulating mast cell function, the effects of ETBE, TBA and ethanol on the levels of intracellular calcium, phosphorylation of Akt (as a marker of phosphatidylinositol 3-kinase) and global tyrosine phosphorylation were also measured as indicators of mast cell activation. In the presence of ETBE, TBA or ethanol, IgE-induced release of beta-hexosaminidase was decreased. These compounds also attenuated the IgE-mediated increase in the levels of intracellular Ca(2+), phosphorylation of Akt and global tyrosine phosphorylation in RBL2H3 cells. ETBE, TBA and ethanol inhibited mast cell degranulation by inhibiting the increase in intracellular calcium ion concentration and activation of phosphatidylinositol 3-kinase and protein tyrosine kinase activation, suggesting that exposure to ETBE might affect immune responses, particularly in allergic diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
tert-Butyl ethyl ether, 99%
Supelco
tert-Butyl ethyl ether, analytical standard
Sigma-Aldrich
tert-Butyl ethyl ether, purum, ≥97.0% (GC)