Merck
  • Acyloxymethyl esterification of nodularin-R and microcystin-LA produces inactive protoxins that become reactivated and produce apoptosis inside intact cells.
Acyloxymethyl esterification of nodularin-R and microcystin-LA produces inactive protoxins that become reactivated and produce apoptosis inside intact cells.
Journal of medicinal chemistry (2009-08-27)
Lars Herfindal, Franciszek Kasprzykowski, Frank Schwede, Leszek Łankiewicz, Kari E Fladmark, Joanna Łukomska, Matti Wahlsten, Kaarina Sivonen, Zbigniew Grzonka, Bernd Jastorff, Stein Ove Døskeland
ABSTRAKT
We report the esterification of the carboxyl groups of the cyclic peptide toxins nodularin-R and microcystin-LA to produce stable diacetoxymethyl and dipropionyloxymethyl ester derivatives. The derivatives had no activity but were reactivated upon esterase treatment. When injected into cells, the acyloxymethyl moieties were cleaved off and apoptosis induced. Linking the acyloxymethyl-ester moiety of these potent toxins to carriers destined for endocytosis paves the way for selective apoptosis induction in target (e.g., cancer) cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phenylglyoxal monohydrate, crystallized, ≥97.0% (GC)