Merck
Squalene emulsions for parenteral vaccine and drug delivery.
Molecules (Basel, Switzerland) (2009-09-29)
Christopher B Fox
ABSTRAKT
Squalene is a linear triterpene that is extensively utilized as a principal component of parenteral emulsions for drug and vaccine delivery. In this review, the chemical structure and sources of squalene are presented. Moreover, the physicochemical and biological properties of squalene-containing emulsions are evaluated in the context of parenteral formulations. Historical and current parenteral emulsion products containing squalene or squalane are discussed. The safety of squalene-based products is also addressed. Finally, analytical techniques for characterization of squalene emulsions are examined.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Squalane, 96%
Sigma-Aldrich
Squalane, technical, ≥90% (GC)
Supelco
Squalene, analytical standard
Sigma-Aldrich
Squalene, ≥98%, liquid