Merck
Essential oil composition of lady's mantle (Alchemilla xanthochlora Rothm.) growing wild in Alpine pastures.
Natural product research (2009-10-08)
Luca Falchero, Mauro Coppa, Alessia Fossi, Giampiero Lombardi, Daniele Ramella, Aldo Tava
ABSTRAKT
The volatile fraction from the aerial part of Alchemilla xanthochlora Rothm. (Rosaceae) was obtained by hydrodistillation, and the chemical composition of the obtained oil was determined by GC/FID and GC/MS. Several compounds were found, belonging to the classes of aldehydes, alcohols, terpenes, esters, acids and hydrocarbons. The major constituents were cis-3-hexenol (11.20+/-0.02%), linalool (10.36+/-0.75%), oct-1-en-3-ol (8.98+/-1.43%), and nonanal (7.83+/-0.93%), followed by myrtenol (4.65+/-0.40%), hexadecanoic acid (4.59+/-0.79%), cis-3-hexenyl acetate (3.92+/-0.15%) and alpha-terpineol (3.10+/-0.26%).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%
Supelco
Nonanal, analytical standard
Sigma-Aldrich
Nonanal, 95%
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Sigma-Aldrich
Nonanal, ≥95%, FCC
Supelco
cis-3-Hexen-1-ol, analytical standard