Merck
Chitin-based organic networks: an integral part of cell wall biosilica in the diatom Thalassiosira pseudonana.
Angewandte Chemie (International ed. in English) (2009-11-20)
Eike Brunner, Patrick Richthammer, Hermann Ehrlich, Silvia Paasch, Paul Simon, Susanne Ueberlein, Karl-Heinz van Pée
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis