Merck
Dissolved core alginate microspheres as "smart-tattoo" glucose sensors.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference (2009-12-08)
Ayesha Chaudhary, Monica Raina, Michael J McShane, Rohit Srivastava
ABSTRAKT
The feasibility of multilayer thin film coated dissolved-core alginate-templated microsphere sensors based on fluorescence resonance energy transfer and competitive binding, was explored in simulated interstitial fluid, using glucose as a model analyte. The glucose sensitivity was observed to be 0.89%/mM glucose with a linear response in the range of 0-50 mM glucose. The sensor response was observed to be completely reversible in nature with a response time of 120 seconds. The system was further demonstrated to respond similarly using near-infrared dyes (Alexa Fluor-647-labeled dextran as donor and QSY-21-conjugated apo-GOx as acceptor) which exhibited a sensitivity of 0.94%/mM glucose with a linear response in range of 0-50 mM glucose, making the sensor more amenable to in vivo use, when implanted in scattering tissue.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
8-(6-Aminohexyl)aminoadenosine 3′:5′-cyclic monophosphate, ~98%