Merck
Synthesis of tetrahydropyrroloiminoquinone alkaloids based on electrochemically generated hypervalent iodine oxidative cyclization.
Organic letters (2009-12-31)
Keisuke Inoue, Yuichi Ishikawa, Shigeru Nishiyama
ABSTRAKT
An approach to the synthesis of the tetrahydropyrroloiminoquinone alkaloids has been developed and applied to the preparation of N-1-beta-D-ribofuranosyltetrahydropyrroloiminoquinones. The strategy utilizes oxidative cyclization of aryl-methoxyamides by hypervalent iodine to construct the quinoline framework shared by members of this alkaloid family. The hypervalent iodine oxidant is generated in situ by anodic oxidation of iodobenzene.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iodobenzene, 98%