Merck
Contamination of rice by etofenprox, diethyl phthalate and alkylphenols: effects on first delivery and sperm count in mice.
The Journal of toxicological sciences (2010-02-02)
Kunioki Hayashi, Ai Nakae, Yukie Fukushima, Kazuya Sakamoto, Tsutomu Furuichi, Kana Kitahara, Yukiko Miyazaki, Chisato Ikenoue, Satoshi Matumoto, Takayoshi Toda
ABSTRAKT
We observed that first delivery was delayed when a group of paired mice fed with non-organic common rice compared to a group fed with organic rice. This led us to hypothesize that pesticides or other soil contaminants may be responsible for the effect on mice reproduction. We then found that the non-organic rice was contaminated with a pesticide etofenprox and nonylphenol, butylphenol and diethyl phthalate which are used as agricultural detergents or plasticizers of agricultural film, that are all suspected to be estrogenic. Therefore, the chemicals were administered to mice at the levels detected in rice, and we subsequently observed that first delivery and sperm count of the animals were significantly impaired. This study is the first to show that rice contaminated with agricultural chemicals affects reproduction in the mammal.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Etofenprox, PESTANAL®, analytical standard