Merck
Bioavailability of diuron, imazapic and isoxaflutole in soils of contrasting textures.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2010-02-26)
Miriam H Inoue, Rubem S Oliveira, Jamil Constantin, Diego G Alonso, Cássio A Tormena
ABSTRAKT
This research was aimed at understanding the dynamics of the herbicides diuron [3-(3,4-dichlorophenyl)-1,1-dimethylurea], imazapic [2-(4-isopropyl-4-methyl-5-oxo-2-imidazolin-2-yl)-5-methylnicotinic acid] and isoxaflutole [5-cyclopropyl-4-(2-methanesulfonyl-4-trifluoromethyl benzoyl)isoxazole] in two soils of different physico-chemical properties. To accomplish such intent, several greenhouse experiments were run. The bioavailability of diuron (0; 1.6 and 3.2 kg ha(-1)), imazapic (0; 98 and 122.5 g ha(-1)) and isoxaflutole (0; 35 and 70 g ha(-1)) was measured in samples from a sandy loam soil and a clay soil, by sowing a bioindicator (Brachiaria decumbens), at 0, 25, 50, 75 and 100 days after herbicides application (DAA). Diuron was very stable in clay soil, providing control equal to or higher than 92% of bioindicator, up to 100 DAA, as assumed by biomass accumulation. No differential effect was observed in sandy loam soil, even when 2x labeled rate were applied. Imazapic provided a short bioavailability in relation to B. decumbens, independent of rates applied. The persistence of isoxaflutole was longer in clay soil (28 to 30 days).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Isoxaflutole, PESTANAL®, analytical standard