Merck
Bioactivity of immobilized EGF on self-assembled monolayers: optimization of the immobilization process.
Journal of biomedical materials research. Part A (2010-03-04)
Raquel Gonçalves, Maria Cristina Lopes Martins, Maria José Oliveira, Graça Almeida-Porada, Mário Adolfo Barbosa
ABSTRAKT
Last trends in Biomaterials focus the mimic of cellular environments capable to control cellular responses. Epidermal growth factor (EGF) is a pleiotropic cytokine known to regulate cell proliferation, differentiation, and death. This study aims to optimize the immobilization of EGF on 11-mercapto-1-undecyl-tetra(ethylene)glycol (EG4)-self-assembled monolayers (SAMs) and to establish a new model surface to study EGF-mediated signaling. Gold substrates were modified with a monolayer of EG4 and N,N'-carbonyldiimidazole (CDI) was used to activate hydroxyl terminated groups of EG4-SAMs. EGF was then immobilized on activated EG4-SAMs at pH 7.4, 4 degrees C, and 100 rpm. Different immobilization reaction times were tested as well as different CDI concentrations to optimize the reaction conditions and obtain a range of immobilized EGF concentrations on the surfaces. Surface characterization of EGF-SAMs was performed using radiolabeling, water contact angle measurements, X-ray photoelectron spectroscopy, and ELISA. Phosphorylation of EGFR on BT-20 breast cancer cell line by EGF-SAMs was tested by immunostaining. EGF was successfully immobilized on EG4-SAMs, at 4 degrees C and pH 7.4 in a range of concentrations from 3.6 +/- 0.8 to 17.6 +/- 1.5 ng/cm(2). The concentration of EGF increases with immobilization time and with the CDI concentration reaching the maximum for surfaces activated with 30 mg/mL of CDI after 48 h. The bioactivity of EGF-SAMs was confirmed by immunostaining of phospho-EGFR of BT-20 cells. This study described EGF immobilization on EG4-SAMs at different concentrations, which could be important surface models to study specific protein interactions at the molecular level evolving EGF-family of proteins.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
CDI, reagent grade