Merck
Efficacy and tolerability of a local acting antiseptic agent in the treatment of vaginal dysbiosis during pregnancy.
Archives of gynecology and obstetrics (2010-03-11)
Volker Briese, Gerd Neumann, Juliane Waldschläger, Theodor W May, Jörg Siebert, Bernd Gerber
ABSTRAKT
In the present study, the efficacy and tolerability of the local acting antiseptic octenidine hydrochloride/phenoxyethanol (OHP) for the treatment of vaginal dysbiosis (VD) and/or bacterial vaginosis (BV) in pregnancy and its potential influence on preterm births and low-weight newborns were examined. One-hundred nine pregnant women with increased pH values (>4.5) and BV characteristic symptoms were treated with OHP for 7 days and a second time in case of a recurrent pH increase. pH values were continuously controlled by women's self-measurements. pH decreased to ≤ 4.5 in 67.9% of patients. Seven of 12 women (58.3%) treated again with OHP due to a recurrent pH increase finally reached the pH target (pH ≤ 4.5). No preterm birth occurred in the OHP group; no newborn had a birth weight <2,000 g. Rates of preterm births and low-weight newborns were comparable between OHP group and pregnant women without VD/BV. OHP is effective and well tolerated in the treatment of VD/BV also in pregnancy without side effects and the occurrence of resistances. It could be an additional therapeutic option in the prevention of the multifactorial disease pattern 'preterm birth' with all their consequences.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%