Merck
Chemical synthesis of heterocyclic-sugar nucleoside analogues.
Chemical reviews (2010-03-18)
Giovanni Romeo, Ugo Chiacchio, Antonino Corsaro, Pedro Merino
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Dioxolane, anhydrous, contains ~75 ppm BHT as inhibitor, 99.8%
Sigma-Aldrich
1,3-Dioxolane, ReagentPlus®, contains ~75 ppm BHT as inhibitor, 99%