Merck
Secondary metabolites of the phytopathogen Peronophythora litchii.
Natural product communications (2010-03-26)
Haihui Xie, Yaoguang Liang, Jinghua Xue, Qiaolin Xu, Yueming Jiang, Xiaoyi Wei
ABSTRAKT
Two new metabolites, 4R*-hydroxy-3,5R*-dimethoxycyclohex-2-enone (1) and 2-hydroxy-N-(2-hydroxyphenyl)propionamide (2), were isolated from solid cultures of the phytopathogen Peronophythora litchii, along with 4-hydroxybenzyl alcohol (3) and methyl D-indole-3-lactate (4). Their structures were elucidated on the basis of spectroscopic and spectrometric data. These metabolites were not toxic to brine shrimps (LC50 > 500 microg/mL).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Hydroxybenzyl alcohol, 99%
Sigma-Aldrich
4-Hydroxybenzyl alcohol, ≥98%, FG