Merck
3-Alkynyl selenophene protects against carbon-tetrachloride-induced and 2-nitropropane-induced hepatic damage in rats.
Cell biology and toxicology (2010-04-17)
Ethel Antunes Wilhelm, Cristiano Ricardo Jesse, Marina Prigol, Diego Alves, Ricardo Frederico Schumacher, Cristina Wayne Nogueira
ABSTRAKT
The aim of this study was to investigate the protective effect of 3-alkynyl selenophene (3-ASP) on acute liver injury induced by carbon tetrachloride (CCl(4)) and 2-nitropropane (2-NP) in rats. On the first day of treatment, the animals received 3-ASP (25 mg/kg, p.o.). On the second day, the rats received CCl(4) (1 mg/kg, i.p.) or 2-NP (100 mg/kg, p.o.). Twenty-four hours after CCl(4) or 2-NP administration, the animals were euthanized, and their plasma and liver were removed for biochemical and histological analyses. The histological analysis revealed extensive injury in the liver of CCl(4)-exposed and 2-NP-exposed rats, which was attenuated by 3-ASP. 3-ASP significantly attenuated (1) the increase in plasmatic aspartate and alanine aminotransferase activities and lipid peroxidation levels induced by CCl(4) and 2-NP; (2) the inhibition of δ-aminolevulinic dehydratase activity caused by 2-NP; and (3) the decrease in ascorbic acid (AA) levels and catalase (CAT) activity caused by CCl(4). AA levels and CAT activity remained unaltered in the liver of rats exposed to 2-NP. The protective effect of 3-ASP on acute liver injury induced by CCl(4) and 2-NP in rats was demonstrated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Nitropropane, ≥96%